Skip to Content
Merck
All Photos(1)

Key Documents

343641

Sigma-Aldrich

(R)-(+)-Citronellal

technical grade

Synonym(s):

(+)-Citronellal, (3R)-3,7-Dimethyl-6-octenal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CHO
CAS Number:
Molecular Weight:
154.25
Beilstein:
1720791
EC Number:
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

form

liquid

refractive index

n20/D 1.448 (lit.)

bp

207 °C (lit.)

density

0.851 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

C[C@H](CC\C=C(\C)C)CC=O

InChI

1S/C10H18O/c1-9(2)5-4-6-10(3)7-8-11/h5,8,10H,4,6-7H2,1-3H3/t10-/m1/s1

InChI key

NEHNMFOYXAPHSD-SNVBAGLBSA-N

General description

(R)-(+)-Citronellal is a monoterpenoid aldehyde. It is a chiral organic compound used as a building block in organic synthesis to prepare various chiral compounds. It is also used as a chiral catalyst or ligand in asymmetric reactions.

Application

(R)-(+)-Citronellal can be used as a reactant to synthesize various compounds such as L-menthol, caparratriene, and chalcogen-containing nitrone derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xiangxian Ying et al.
Molecules (Basel, Switzerland), 24(6) (2019-03-21)
The members of the Old Yellow Enzyme (OYE) family are capable of catalyzing the asymmetric reduction of (E/Z)-citral to (R)-citronellal-a key intermediate in the synthesis of L-menthol. The applications of OYE-mediated biotransformation are usually hampered by its insufficient enantioselectivity and
Walter S Leal et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(46), 18704-18709 (2013-10-30)
The southern house mosquito, Culex quinquefasciatus, has one of the most acute and eclectic olfactory systems of all mosquito species hitherto studied. Here, we used Illumina sequencing to identify olfactory genes expressed predominantly in antenna, mosquito's main olfactory organ. Less
Lucindo José Quintans-Júnior et al.
Journal of orofacial pain, 24(3), 305-312 (2010-07-29)
To evaluate the antinociceptive effects of citronellal (CTL) on formalin-, capsaicin-, and glutamate-induced orofacial nociception in mice and to investigate whether such effects might involve a change in neural excitability. Male mice were pretreated with CTL (50, 100, and 200
Marisa Belicchi-Ferrari et al.
Journal of inorganic biochemistry, 104(2), 199-206 (2009-12-08)
This paper reports the syntheses and characterization of ethylmorpholine substituted citronellal thiosemicarbazone copper(II) and nickel(II) metal complexes. The compounds were characterized through elemental analyses and spectroscopic (IR, UV-Vis, NMR, MS) methods. The X-ray analysis of the two complexes shows that
M S Melo et al.
Pharmaceutical biology, 48(4), 411-416 (2010-07-22)
Citronellal is a monoterpene reported to be a major component of the essential oils in various aromatic species of plants. The present study evaluated the central nervous system depressant and antinociceptive properties of citronellal through behavioral experimental models. Following intraperitoneal

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service