Skip to Content
Merck
All Photos(1)

Key Documents

341576

Sigma-Aldrich

(3aR,4S,5R,6aS)-(−)-Hexahydro-5-hydroxy-4-(hydroxymethyl)-2H-cyclopenta[b]furan-2-one

98%

Synonym(s):

(−)-Corey lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O4
CAS Number:
Molecular Weight:
172.18
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

optical activity

[α]20/D −44°, c = 1.4 in methanol

mp

117-119 °C (lit.)

functional group

ester
hydroxyl

SMILES string

OC[C@H]1[C@H](O)C[C@@H]2OC(=O)C[C@H]12

InChI

1S/C8H12O4/c9-3-5-4-1-8(11)12-7(4)2-6(5)10/h4-7,9-10H,1-3H2/t4-,5-,6-,7+/m1/s1

InChI key

VYTZWRCSPHQSFX-GBNDHIKLSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erin M Anderson et al.
Tissue engineering. Part A, 21(7-8), 1217-1227 (2014-12-02)
Biomaterials may improve outcomes of endothelial progenitor-based therapies for the treatment of ischemic cardiovascular disease, due to their ability to direct cell behavior. We hypothesized that local, sustained delivery of exogenous vascular endothelial growth factor (VEGF) and stromal cell-derived factor
Darya Zibrova et al.
The Biochemical journal, 474(6), 983-1001 (2016-12-23)
Activation of AMP-activated protein kinase (AMPK) in endothelial cells regulates energy homeostasis, stress protection and angiogenesis, but the underlying mechanisms are incompletely understood. Using a label-free phosphoproteomic analysis, we identified glutamine:fructose-6-phosphate amidotransferase 1 (GFAT1) as an AMPK substrate. GFAT1 is
Jayeeta Ghosh et al.
Polymer, 52(12), 2650-2660 (2011-07-23)
The objective of this research was to examine the capabilities of QSPR (Quantitative Structure Property Relationship) modeling to predict specific biological responses (fibrinogen adsorption, cell attachment and cell proliferation index) on thin films of different polymethacrylates. Using 33 commercially available
Aaron J Rogers et al.
Cells, tissues, organs, 206(1-2), 82-94 (2019-03-07)
Human induced pluripotent stem cell (hiPSC)-derived cardio-myocytes (hiPSC-CMs) hold great promise for cardiovascular disease modeling and regenerative medicine. However, these cells are both structurally and functionally -immature, primarily due to their differentiation into cardiomyocytes occurring under static culture which only
Guillaume Beauclair et al.
Journal of virology, 94(5) (2019-12-13)
Kaposi's sarcoma-associated herpesvirus (KSHV) is the cause of three human malignancies: Kaposi's sarcoma, primary effusion lymphoma, and the plasma cell variant of multicentric Castleman disease. Previous research has shown that several cellular tyrosine kinases play crucial roles during several steps

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service