Skip to Content
Merck
All Photos(2)

Documents

339237

Sigma-Aldrich

Zirconium(IV) isopropoxide isopropanol complex

99.9% trace metals basis

Synonym(s):

Propan-2-ol propan-2-olate zirconium(4+), Zirconium (2-propanol)tetrakis(2-propanolato)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zr(OCH(CH3)2)4 · (CH3)2CHOH
CAS Number:
Molecular Weight:
387.67
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

core: zirconium

SMILES string

CC(C)O.CC(C)O[Zr](OC(C)C)(OC(C)C)OC(C)C

InChI

1S/C3H8O.4C3H7O.Zr/c5*1-3(2)4;/h3-4H,1-2H3;4*3H,1-2H3;/q;4*-1;+4

InChI key

NIJDLRJGRCHJDB-UHFFFAOYSA-N

Application

Convenient precursor for the synthesis of olefin-functionalized zirconium-containing monomers, which have potential for the generation of hybrid zirconium-containing polymers.
Zirconium(IV) isopropoxide isopropanol complex may be used as a precursor to ZrO2.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and properties of visible light responsive ZrTiO 4/Bi 2 O 3 photocatalysts for 4-chlorophenol decomposition.
Neppolian B, et al.
Journal of Hazardous Materials, 182(1), 557- 562 (2010)
Synthesis and characterization of ZrO 2?TiO 2 binary oxide semiconductor nanoparticles: application and interparticle electron transfer process.
Neppolian B, et al.
Applied Catalysis A: General, 333(2), 264-271 (2007)
William J. Evans et al.
Inorganic chemistry, 38(6), 1160-1164 (2001-10-24)
The attachment of pendent olefin groups to oxygen-ligated zirconium complexes using olefin-substituted phenols and alcohols and readily accessible zirconium reagents is described. Syntheses of three crystallographically characterizable complexes isolable in 55-90% yield are reported. Eugenol, HOC(6)H(3)(OMe-2)(CH(2)CH=CH(2)-4) (HOAr) reacts with [Zr(O(i)Pr)(4)(HO(i)Pr)](2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service