Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
[Ru(COD)Cl2]n
CAS Number:
Molecular Weight:
280.16 (as monomer)
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22
Quality Level
assay
95%
reaction suitability
core: ruthenium, reaction type: solution phase peptide synthesis, reagent type: catalyst
greener alternative product characteristics
Catalysis
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
greener alternative category
, Aligned
SMILES string
[Ru+2].[Cl-].[Cl-].C1CC=CCCC=C1
InChI
1S/C8H12.2ClH.Ru/c1-2-4-6-8-7-5-3-1;;;/h1-2,7-8H,3-6H2;2*1H;/q;;;+2/p-2
InChI key
DMRVBCXRFYZCPR-UHFFFAOYSA-L
General description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.
Application
Versatile Ru catalyst precursor. Used with IMes to catalyze the dehydrogenative coupling of alcohols and amines to form amide bonds.
Versatile ruthenium catalyst precursor. Used with NHC ligand to catalyze greener amide bond formation by dehydrogenative coupling of amines and alcohols.
Amide Synthesis from Alcohols and Amines by the Extrusion of Dihydrogen
Amide Synthesis from Alcohols and Amines by the Extrusion of Dihydrogen
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 337331-1G | 04061826743539 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service