Skip to Content
Merck
All Photos(1)

Key Documents

334677

Sigma-Aldrich

2-Imino-4-thiobiuret

99%

Synonym(s):

Amidinothiourea

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC(=NH)NHCSNH2
CAS Number:
Molecular Weight:
118.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

171-173 °C (lit.)

functional group

amine
thiourea

SMILES string

NC(=N)NC(N)=S

InChI

1S/C2H6N4S/c3-1(4)6-2(5)7/h(H6,3,4,5,6,7)

InChI key

OKGXJRGLYVRVNE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

[Prevention of postresuscitation heart damage with gutimine].
V T Dolgikh
Anesteziologiia i reanimatologiia, (1)(1), 59-62 (1987-01-01)
Metal complexes used as Anti-Inflammatory agents: Synthesis, characterization and anti-inflammatory action VO (II)-complexes.
Shukla S and Mishra AP
Arabian Journal of Chemistry (2014)
N Iu Semigolovskiĭ et al.
Ontogenez, 19(1), 89-93 (1988-01-01)
A one-time addition of gutimine (0.5 g/l), amtizol (0.2 g/l), and unithiol (0.25 g/l) to the medium at early stages of tadpole development was shown to inhibit metamorphosis only upon addition of unithiol. Repeated addition of gutimine, amtizol, and unithiol
G A Boiarinov et al.
Voprosy meditsinskoi khimii, 39(4), 34-38 (1993-07-01)
Physico-chemical properties of phospholipids were studied in heart muscle at postischemic period after preventive treatment with gutimine. Two administrations of the drug, as compared with its three times treatment, during the preischemic period contributed to development of stable adaptive antiischemic
[Hypoxia and its pharmacological correction: a key problem of anesthesiology and intensive care].
A V Smirnov et al.
Anesteziologiia i reanimatologiia, (3)(3), 97-98 (1997-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service