Skip to Content
Merck
All Photos(2)

Key Documents

328510

Sigma-Aldrich

4′-Pentyl-4-biphenylcarbonitrile

liquid crystal (nematic), 98%

Synonym(s):

4-Cyano-4-pentylbiphenyl, 4-Pentyl-4-cyanobiphenyl, 5CB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4C6H4C6H4CN
CAS Number:
Molecular Weight:
249.35
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid crystal (nematic)

refractive index

n20/D 1.532 (lit.)

bp

140-150 °C/0.5 mmHg (lit.)

density

1.008 g/mL at 25 °C (lit.)

SMILES string

CCCCCc1ccc(cc1)-c2ccc(cc2)C#N

InChI

1S/C18H19N/c1-2-3-4-5-15-6-10-17(11-7-15)18-12-8-16(14-19)9-13-18/h6-13H,2-5H2,1H3

InChI key

HHPCNRKYVYWYAU-UHFFFAOYSA-N

General description

4′-Pentyl-4-biphenylcarbonitrile (5CB) is a nematic liquid crystal. 5CB undergoes phase transition from crystalline to nematic then to isotropic state at 18oC and 35oC respectively. It may be produced by modifying biphenyl.2

Application

Attaching 5CB to an aqueous-liquid crystal interface by redox active surfactant has been investigated.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Pan et al.
Nanomaterials (Basel, Switzerland), 8(6) (2018-06-13)
Resistive pressure sensors generally employ microstructures such as pores and pyramids in the active layer or on the electrodes to reduce the Young’s modulus and improve the sensitivity. However, such pressure sensors always exhibit complex fabrication process and have difficulties
Yu Xia et al.
Advanced materials (Deerfield Beach, Fla.), 28(43), 9637-9643 (2016-10-08)
Two-dimensional liquid-crystal elastomer (LCE) sheets with preprogrammed topological defects are prepared by aligning liquid-crystal monomers within micropatterned epoxy channels, followed by photopolymerization. Upon heating, the LCE films form various three-dimensional structures in agreement with theoretical design. The miniaturized LCE actuators
Makiko T Quint et al.
Scientific reports, 7(1), 17788-17788 (2017-12-21)
We present three-dimensional microshells formed by self-assembly of densely-packed 5 nm gold nanoparticles (AuNPs). Surface functionalization of the AuNPs with custom-designed mesogenic molecules drives the formation of a stable and rigid shell wall, and these unique structures allow encapsulation of cargo
Ahlam Nemati et al.
ACS nano, 13(9), 10312-10326 (2019-08-20)
The creation and transmission of chirality in molecular systems is a well-known, widely applied notion. Our understanding of how the chirality of nanomaterials can be controlled, measured, transmitted through space, and applied is less well understood. Dynamic assemblies for chiral
Jeffrey M Brake et al.
Langmuir : the ACS journal of surfaces and colloids, 23(16), 8497-8507 (2007-06-28)
We report a study of the interactions of proteins with monolayers of phospholipids (D/L-alpha-dipalmitoyl phosphatidylcholine and L-alpha-dilauroyl phosphatidylcholine) spontaneously assembled at an interface between an aqueous phase and a 20-microm-thick film of a nematic liquid crystal (4'-pentyl-4-cyanobiphenyl). Because the orientation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service