Skip to Content
Merck
All Photos(4)

Documents

317020

Sigma-Aldrich

(−)-DIP-Chloride

Synonym(s):

(−)-B-Chlorodiisopinocampheylborane, (−)-Ipc2BCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34BCl
CAS Number:
Molecular Weight:
320.75
Beilstein:
5937756
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

52-56 °C (lit.)

SMILES string

C[C@H]1[C@@H](CC2CCC1C2(C)C)B(Cl)[C@@H]3CC4CCC([C@H]3C)C4(C)C

InChI

1S/C20H34BCl/c1-11-15-7-13(19(15,3)4)9-17(11)21(22)18-10-14-8-16(12(18)2)20(14,5)6/h11-18H,7-10H2,1-6H3/t11-,12-,13+,14+,15-,16-,17-,18-/m1/s1

InChI key

PSEHHVRCDVOTID-VMAIWCPRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Both (+)- and (-)-DIP-chloride are used for asymmetric reduction of prochiral ketones and for the preparation of β-amino alcohols.
Reagent used for an enantioselective reduction of (chloroacetyl) benzofurans to chlorohydrins which serve as intermediates for the corresponding epoxides and aminoalcohols. For a short paper on the preparation of DIP halides and their use in stereoselective reductions see Chem. Commun. 1053 (1994).

Legal Information

DIP-Chloride is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Asymmetry, 16, 3205-3205 (2005)
Katie M Cergol et al.
Nature protocols, 2(10), 2568-2573 (2007-10-20)
The protocol for the preparation of boron enolates and their subsequent reaction with aldehydes is described, providing convenient access to beta-hydroxy ketones in good yields and with high stereoselectivities. The reaction consists of three steps: first, the ketone is rapidly
Bull. Korean Chem. Soc., 26, 652-652 (2005)
Journal of the Chemical Society. Chemical Communications, 1053-1053 (1994)
Tetrahedron Letters, 35, 1511-1511 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service