Skip to Content
Merck
All Photos(1)

Key Documents

311308

Sigma-Aldrich

1,4,7-Triazacyclononane

95%

Synonym(s):

Octahydro-1H-1,4,7-triazonine, Triethylenetriamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15N3
CAS Number:
Molecular Weight:
129.20
Beilstein:
773877
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

bp

110-130 °C/7 mmHg (lit.)

mp

42-45 °C (lit.)

SMILES string

C1CNCCNCCN1

InChI

1S/C6H15N3/c1-2-8-5-6-9-4-3-7-1/h7-9H,1-6H2

InChI key

ITWBWJFEJCHKSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Possible reagent for compleximetric titrations with high cation-binding selectivity.
Starting material for the synthesis of 1,4,7-trifunctionalized derivatives which have applications in metal complexation.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Journal of the Chemical Society. Chemical Communications, 1473-1473 (1991)
Didar Asik et al.
Molecules (Basel, Switzerland), 25(10) (2020-05-18)
Complexes of Fe(III) that contain a triazacyclononane (TACN) macrocycle, two pendant hydroxyl groups, and a third ancillary pendant show promise as MRI contrast agents. The ancillary group plays an important role in tuning the solution relaxivity of the Fe(III) complex
Jun-Fang Guo et al.
Inorganic chemistry, 49(4), 1607-1614 (2010-01-14)
A novel dicyanoosmium(III) complex, trans-Ph(4)P[Os(III)(salen)(CN)(2)].CH(2)Cl(2).H(2)O (1; Ph(4)P(+) = tetraphenylphosphonium cation, salen(2-) = N,N'-ethylenebis(salicylideneaminato) dianion), has been synthesized and structurally characterized. Reactions of 1 with [Cu(Me(3)tacn)(H(2)O)(2)](ClO(4))(2) (Me(3)tacn = 1,4,7-trimethyl-1,4,7-triazacyclononane) under different conditions produce the 1-D ferromagnetic zigzag chains [Os(salen)(CN)(2)](2)[Cu(Me(3)tacn)].CH(3)OH (2) and
Aline Nonat et al.
Inorganic chemistry, 48(9), 4207-4218 (2009-04-08)
The tripodal H(3)thqtcn ligand allows the synthesis of well-defined neutral monomeric syn-tris(hydroxyquinolinate) complexes of lanthanides. Pure [Ln(thqtcn)] complexes (Ln = Nd, 1; Er, 2; Yb, 3) of the triply deprotonated ligand thqtcn(3-) were prepared. Crystallographic characterization was carried out for
Taizo Mori et al.
Journal of the American Chemical Society, 132(37), 12868-12870 (2010-09-03)
Construction of enzyme-like artificial cavities is a complex and challenging subject. Rather than synthesizing complicated host molecules, we have proposed mechanical adaptation of relatively simple hosts within dynamic media to determine the optimum conformation for molecular recognition. Here we have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service