Skip to Content
Merck
All Photos(2)

Documents

311278

Sigma-Aldrich

5-Hydroxyisophthalic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3-1,3-(CO2H)2
CAS Number:
Molecular Weight:
182.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

298-302 °C (lit.)

SMILES string

OC(=O)c1cc(O)cc(c1)C(O)=O

InChI

1S/C8H6O5/c9-6-2-4(7(10)11)1-5(3-6)8(12)13/h1-3,9H,(H,10,11)(H,12,13)

InChI key

QNVNLUSHGRBCLO-UHFFFAOYSA-N

General description

The hydrothermal reaction of 5-hydroxyisophthalic acid, Co(II)/Cu(II) and dipyridophenazine, that forms two 3D chiral coordination polymers, was studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mamoru Yamada et al.
Biotechnology letters, 32(3), 445-450 (2009-11-27)
5-Hydroxyisophthalic acid-producing microorganisms were isolated from enrichment cultures using 5-sulfoisophthalic acid as a sulfur source. One bacterium, Ochrobactrum anthropi S9, had the highest 5-sulfoisophthalic acid-degrading activity, and stoichiometrically formed 5-hydroxyisophthalic acid, a raw material for polymer synthesis. Under optimum culture
Zheng-Bo Han et al.
Dalton transactions (Cambridge, England : 2003), (44)(44), 9807-9811 (2009-11-04)
The hydrothermal reaction of Co(II)/Cu(II), 5-hydroxyisophthalic acid and dipyridophenazine leads to the generation of two 3D chiral coordination polymers, [M(hip)(DPPZ)](n) (M = Co(), Cu(), H(2)hip = 5-hydroxyisophthalic acid, DPPZ = dipyridophenazine), which contain M-hip-M helical chains (M = Co, Cu)
Jiyoung Lee et al.
Nature communications, 8, 14070-14070 (2017-01-05)
Self-assembly has proven to be a widely successful synthetic strategy for functional materials, especially for metal-organic materials (MOMs), an emerging class of porous materials consisting of metal-organic frameworks (MOFs) and metal-organic polyhedra (MOPs). However, there are areas in MOM synthesis
Kaisa Hänninen et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 31(5), 306-317 (2007-06-22)
Donnan theory was applied to gain mechanistic understanding on the factors affecting drug loading process, compound-fiber affinity and subsequent release from fibrous ion-exchangers. Impact of initial loading solution concentration on fiber occupancy and loading efficiency of compounds were assessed experimentally
Mathias Köberl et al.
Dalton transactions (Cambridge, England : 2003), 40(43), 11490-11496 (2011-09-29)
The treatment of the dimeric paddle-wheel (PW) compound [Mo(2)(NCCH(3))(10)][BF(4)](4)1 with oxalic acid (0.5 equiv.), 1,1-cyclobutanedicarboxylic acid (1 equiv.), 5-hydroxyisophthalic acid (1 equiv.) (m-bdc-OH) or 2,3,5,6-tetrafluoroterephthalic acid (0.5 or 1 equiv.) leads to the formation of macromolecular dicarboxylate-linked (Mo(2))(n) entities (n

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service