Skip to Content
Merck
All Photos(1)

Key Documents

308080

Sigma-Aldrich

Benzoylnitromethane

98%

Synonym(s):

α-Nitroacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH2NO2
CAS Number:
Molecular Weight:
165.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

105-107 °C (lit.)

storage temp.

2-8°C

SMILES string

[O-][N+](=O)CC(=O)c1ccccc1

InChI

1S/C8H7NO3/c10-8(6-9(11)12)7-4-2-1-3-5-7/h1-5H,6H2

InChI key

JTWHVBNYYWFXSI-UHFFFAOYSA-N

Related Categories

General description

The kinetics of proton transfer from benzoylnitromethane to various bases was studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claude F. Bernasconi et al.
The Journal of organic chemistry, 62(23), 8162-8170 (2001-10-24)
The replacement of a hydrogen in nitromethane and in phenylnitromethane by the PhCO group has a strong acidifying effect, i.e., PhCOCH(2)NO(2), 5, is 5.8, 6.6, and 8.6 pK(a) units more acidic than CH(3)NO(2) in water, 50% DMSO-50% water (v/v), and
Parham Taslimi
Archiv der Pharmazie, 353(11), e2000210-e2000210 (2020-09-03)
In this study, the acetophenone derivatives 1-6 were found to be effective inhibitor molecules for α-glycosidase, human carbonic anhydrases I and II (hCA I/II), and acetylcholinesterase (AChE), with Ki values in the range of 167.98 ± 25.06 to 304.36 ± 65.45 µM for α-glycosidase, 555.76 ± 56.07

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service