Skip to Content
Merck
All Photos(1)

Key Documents

303135

Sigma-Aldrich

Triton N-101, reduced

Synonym(s):

Polyoxyethylene branched nonylcyclohexyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H68O10.5
CAS Number:
Molecular Weight:
596.85
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

viscous liquid

Quality Level

refractive index

n20/D 1.468

SMILES string

CC(C)(C)CCC(C)(C)C1CCC(CC1)OCCOCCOCCOCCOCCOCCOCCOCCOCCO.CC(C)(C)CCC(C)(C)C2CCC(CC2)OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C35H70O11.C33H66O10/c1-34(2,3)10-11-35(4,5)32-6-8-33(9-7-32)46-31-30-45-29-28-44-27-26-43-25-24-42-23-22-41-21-20-40-19-18-39-17-16-38-15-14-37-13-12-36;1-32(2,3)10-11-33(4,5)30-6-8-31(9-7-30)43-29-28-42-27-26-41-25-24-40-23-22-39-21-20-38-19-18-37-17-16-36-15-14-35-13-12-34/h32-33,36H,6-31H2,1-5H3;30-31,34H,6-29H2,1-5H3

InChI key

FLXVMMNBMSGKSP-UHFFFAOYSA-N

Application

Triton N-101, reduced has been used:
  • as a non-ionic detergent in phosphate buffer to measure total cytochrome P450 in the biological sample
  • as a component of sucrose solution for the isolation of nuclei from Xenopus embryos
  • as a non-ionic surfactant to study its effects on phenanthrene biodegradation

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Sun Cho et al.
Cold Spring Harbor protocols, 2019(4) (2018-08-23)
Transcriptional regulatory elements are typically found in relatively nucleosome-free genomic regions, often referred to as "open chromatin." Deoxyribonuclease I (DNase I) can digest nucleosome-depleted DNA (presumably bound by transcription factors), but DNA in nucleosomes or higher-order chromatin fibers is less
Synthesis and Physicochemical Properties of Yttrium Oxide Doped with Neodymium and Lanthanum
Kruk A, et al.
Journal of Electronic Materials, 43(9) (2014)
S Y Yuan et al.
Chemosphere, 41(9), 1463-1468 (2000-11-01)
We investigated the potential biodegradation of polycyclic aromatic hydrocarbons (PAHs) by an aerobic mixed culture utilizing phenanthrene as its carbon source. Following a 3-5 h post-treatment lag phase, complete degradation of 5 mg/l phenanthrene occurred within 28 h (optimal conditions
F Peter Guengerich et al.
Nature protocols, 4(9), 1245-1251 (2009-08-08)
Cytochrome P450 (P450) enzymes are important in the metabolism of steroids, vitamins, carcinogens, drugs and other compounds. Two of the commonly used assays in this field are the measurements of total P450 and NADPH-P450 reductase in biological preparations. A detailed
Influence of yttrium oxide on the phase formation and sintering of ceramics based on the MgAl2O4 spinel
Belousova OL, et al.
Glass Phys. Chem., 34(4), 474-479 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service