Skip to Content
Merck
All Photos(1)

Key Documents

297852

Sigma-Aldrich

Ethyl p-tolylacetate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH2CO2C2H5
CAS Number:
Molecular Weight:
178.23
Beilstein:
2045927
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.496 (lit.)

bp

116-118 °C/13 mmHg (lit.)

density

1.008 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)Cc1ccc(C)cc1

InChI

1S/C11H14O2/c1-3-13-11(12)8-10-6-4-9(2)5-7-10/h4-7H,3,8H2,1-2H3

InChI key

BTRGZBIXPLFVNK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl p-tolylacetate was used in preparation of:
  • ethyl-substituted phenylpropioloylacetates
  • ethyl 2-(4-bromomethylphenyl)propionate
  • (−)-incrustoporin, antifungal agent

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 2-(4-bromomethylphenyl) propionic acid and ester.
Liu Z-X and Cheng Q-R.
Chemical Reagents, 4, 021-021 (2011)
The condensation of acetylenic esters with ethyl phenylacetates.
Al-Jallo HN and Al-Hajjar FH.
J. Chem. Soc. Sect. C, 15, 2056-2058 (1970)
Ring-Closing Metathesis Enabled Efficient Synthesis of ?-Butenolide Antifungal Agent (-)-Incrustoporin and its Analogues.
Fernandes RA, et al.
Asian Journal of Organic Chemistry, 3(1), 58-62 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service