Skip to Content
Merck
All Photos(1)

Documents

294519

Sigma-Aldrich

Chloromethyl octyl ether

95%

Synonym(s):

1-(Chloromethoxy)octane, Chloromethyl n-octyl ether, Octyloxymethyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OCH2Cl
CAS Number:
Molecular Weight:
178.70
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

contains

0.1% 2,6-di-tert-butylphenol as stabilizer

refractive index

n20/D 1.437 (lit.)

density

0.924 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCCOCCl

InChI

1S/C9H19ClO/c1-2-3-4-5-6-7-8-11-9-10/h2-9H2,1H3

InChI key

YUMNZEWYPUBSQA-UHFFFAOYSA-N

Application

Chloromethyl octyl ether has been employed as chloromethylation reagent:
  • in chloromethylation of calix[4]arene, methyl ethers of calix[6]arene and calix[8]arene
  • in the synthesis of chloromethylated poly(phthalazinone ether sulfone ketone)
  • in chloromethylation of polyetherethersulphone-polyethersulphone copolymers
  • for polymers

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Polymer Science, 23, 231-231 (1985)
Br. Polym. J., 16, 234-234 (1984)
Synthesis and characterization of quaternized poly (phthalazinone ether sulfone ketone) for anion-exchange membrane.
Jian XG, et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 18(10), 1269-1272 (2007)
Chloromethylation of calixarenes and synthesis of new water soluble macrocyclic hosts.
Almi m, et al.
Tetrahedron, 45(7), 2177-2182 (1989)
On the chloromethylation of polyetherethersulphone/polyethersulphone copolymers. The use of triflic acid as catalyst.
Naik HA, et al.
Polymer, 33(1), 166-169 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service