Skip to Content
Merck
All Photos(1)

Key Documents

29252

Sigma-Aldrich

Cyclohexene-1-carbonitrile

≥97.0%

Synonym(s):

1-Cyanocyclohexene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N
CAS Number:
Molecular Weight:
107.15
Beilstein:
385837
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

refractive index

n20/D 1.482 (lit.)

density

0.946 g/mL at 20 °C (lit.)

functional group

nitrile

SMILES string

N#CC1=CCCCC1

InChI

1S/C7H9N/c8-6-7-4-2-1-3-5-7/h4H,1-3,5H2

InChI key

GTMWGXABXQTZRJ-UHFFFAOYSA-N

General description

Irradiation of cyclohexene-1-carbonitrile with the full arc of Hanovia 450-W lamp yields bicycle[3.1.0]hexane-1-carbonitrile. Photochemistry of cyclohexene-1-carbonitrile has been investigated.

Application

Cyclohexene-1-carbonitrile has been used in the preparation of cyclohexene-1-carboxylic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and photochemistry of cyclohexene-1-carbonitriles.
McCullough JJ and Manning C.
The Journal of Organic Chemistry, 43(14), 2839-2842 (1978)
Aromatization of cyclohexanecarboxylic acid.
B M Babior et al.
The Journal of biological chemistry, 241(16), 3643-3651 (1966-08-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service