Skip to Content
Merck
All Photos(1)

Documents

290998

Sigma-Aldrich

Pentylmagnesium bromide solution

2.0 M in diethyl ether

Synonym(s):

1-Pentylmagnesium bromide, n-Pentylmagnesium bromide, Amylmagnesium bromide, Bromopentylmagnesium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H11MgBr
CAS Number:
Molecular Weight:
175.35
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

2.0 M in diethyl ether

density

0.947 g/mL at 25 °C

SMILES string

CCCCC[Mg]Br

InChI

1S/C5H11.BrH.Mg/c1-3-5-4-2;;/h1,3-5H2,2H3;1H;/q;;+1/p-1

InChI key

ZXWGTPYWPZGZPT-UHFFFAOYSA-M

Application

Pentylmagnesium bromide solution is a Grignard reagent that can be used in the preparation of:
  • Alkanes via Ni or Cu-catalyzed cross-coupling reaction with alkyl fluorides.
  • Hydroxypiperidinones via the addition-cyclization-deprotection process of aldimines.

It can also be used as a reagent to synthesize a key intermediate by olefin alkylation in the total synthesis of alkaloid, (+)-hyperaspine , and surface alkylation of H-terminated Si (111), by Si-C bond formation.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ronia Chung-Tin Sham et al.
Environment international, 137, 105567-105567 (2020-02-23)
The occurrence of triphenyltin (TPT) compounds, a highly toxic antifouling biocide, has been documented in marine environments and organisms all over the world. While some studies showed that marine mammals can be used as sentinel organisms to evaluate the pollution
A six-step asymmetric synthesis of (+)-hyperaspine
Comins DL and Sahn JJ
Organic Letters, 7(23), 5227-5228 (2005)
Diastereoconvergent Synthesis of trans-5-Hydroxy-6-Substituted-2-Piperidinones by Addition-Cyclization-Deprotection Process
Si Chang-Mei, et al.
Organic Letters, 16(16), 4328-4331 (2014)
Alkylation of Si surfaces using a two-step halogenation/Grignard route
Bansal A, et al.
Journal of the American Chemical Society, 118(30), 7225-7226 (1996)
Ni-or Cu-catalyzed cross-coupling reaction of alkyl fluorides with Grignard reagents
Terao J, et al.
Journal of the American Chemical Society, 125(19), 5646-5647 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service