Skip to Content
Merck

288985

2-Thiophenecarbonyl chloride

97%

Synonym(s):

α-Thenoyl chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H3ClOS
CAS Number:
Molecular Weight:
146.59
UNSPSC Code:
12352100
NACRES:
NA.22
PubChem Substance ID:
EC Number:
226-092-4
Beilstein/REAXYS Number:
110145
MDL number:
Assay:
97%
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

97%

form

liquid

refractive index

n20/D 1.590 (lit.)

bp

206-208 °C (lit.)

density

1.371 g/mL at 25 °C (lit.)

functional group

acyl chloride

storage temp.

2-8°C

SMILES string

ClC(=O)c1cccs1

InChI

1S/C5H3ClOS/c6-5(7)4-2-1-3-8-4/h1-3H

InChI key

QIQITDHWZYEEPA-UHFFFAOYSA-N

General description

The postfunctionalization reaction of 2-thiophenecarbonyl chloride with single walled carbon nanotubes (SWCNTs) was studied.

Application

2-Thiophenecarbonyl chloride was used in the synthesis of building blocks derived from ornithine by undergoing acylation/sulphonation of copper complex of orthinine.


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

194.0 °F

flash_point_c

90 °C

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library