Skip to Content
Merck
All Photos(2)

Documents

288225

Sigma-Aldrich

Chloro(triethylphosphine)gold(I)

97%

Synonym(s):

(Triethylphosphine)gold(I) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3PAuCl
CAS Number:
Molecular Weight:
350.58
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

core: gold
reagent type: catalyst

bp

210 °C/0.03 mmHg (lit.)

mp

84-86 °C (lit.)

SMILES string

Cl[Au].CCP(CC)CC

InChI

1S/C6H15P.Au.ClH/c1-4-7(5-2)6-3;;/h4-6H2,1-3H3;;1H/q;+1;/p-1

InChI key

SYBBXLKWGHAVHP-UHFFFAOYSA-M

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A A Isab et al.
Journal of inorganic biochemistry, 38(2), 95-100 (1990-02-01)
New compounds having the formula [L-Au-PEt3]Cl (where L = imidazolidine-2-thione(Imt) and 1,3-Diazinane-2-thione(Diaz)) are synthesized. They are characterized by elemental analysis, infrared, 13C and 31P NMR spectroscopy.
G F Rush et al.
Toxicology and applied pharmacology, 90(3), 377-390 (1987-09-30)
Triethylphosphine gold complexes are effective therapeutic agents used for the treatment of rheumatoid arthritis. Many of those molecules are also highly cytotoxic in vitro and can inhibit DNA and protein synthesis. Preliminary experiments have indicated that triethylphosphine gold chloride (TEPAu)
C E Blank et al.
Journal of inorganic biochemistry, 21(1), 21-29 (1984-05-01)
The DNA binding properties of the Au(I)-containing antiarthritic drug Ridaura and related Au(I) complexes have been investigated. Absorption and circular dichroism spectroscopy show that if the gold ion possesses an easily displaced ligand, e.g., Cl- or Br-, Au(I) is capable
C J Kirkpatrick et al.
Rheumatology international, 3(2), 49-56 (1983-01-01)
Chondrocyte structure and function under the influence of two gold-containing therapeutic agents, aurothioglucose and triethylphosphine gold, were studied in a monolayer culture system for cultivating lapine articular chondrocytes. The functional parameters investigated were chondrocyte proliferation and the incorporation of 35SO4
P Davis et al.
Arthritis and rheumatism, 26(1), 82-86 (1983-01-01)
The effect of sodium aurothiomalate and triethylphosphine gold on the generation of superoxide radicals by chemotactic tripeptide-activated polymorphonuclear leukocytes has been investigated using a cytochrome C reduction technique. Neither gold compound inhibited the binding of the tripeptide to cells. Sodium

Articles

We are proud to offer a treasure-trove of gold precatalysts and silver salts, as well as an extensive portfolio of unsaturated building blocks to accelerate your research success in this exciting field.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service