Skip to Content
Merck
All Photos(1)

Key Documents

286427

Sigma-Aldrich

L-Leucinamide hydrochloride

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(NH2)CONH2·HCl
CAS Number:
Molecular Weight:
166.65
Beilstein:
4237021
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

optical activity

[α]25/D +10°, c = 5 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

254-256 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cl.CC(C)C[C@H](N)C(N)=O

InChI

1S/C6H14N2O.ClH/c1-4(2)3-5(7)6(8)9;/h4-5H,3,7H2,1-2H3,(H2,8,9);1H/t5-;/m0./s1

InChI key

VSPSRRBIXFUMOU-JEDNCBNOSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew R Conrad et al.
The journal of physical chemistry. A, 115(34), 9676-9681 (2011-05-12)
Rotational spectra were recorded for two isotopic species of two conformers of the amide derivative of leucine in the range of 10.5-21 GHz and fit to a rigid rotor Hamiltonian. Ab initio calculations at the MP2/6-311++G(d,p) level identified the low
A conductometric method for the assay of amidase and peptidase activities.
C R Hill et al.
Analytical biochemistry, 120(1), 165-175 (1982-02-01)
[Leucinamide specific cytosol leucine aminopeptidase].
T Kanno
Rinsho byori. The Japanese journal of clinical pathology, 30(5), 502-506 (1982-05-01)
Determination of the enantiomers of ketoprofen in blood plasma by ion-pair extraction and high-performance liquid chromatography of leucinamide derivatives.
S Björkman
Journal of chromatography, 414(2), 465-471 (1987-03-06)
Formation of diastereomeric derivatives of 2-arylpropionic acids using L-leucinamide.
H Spahn
Journal of chromatography, 423, 334-339 (1987-12-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service