Skip to Content
Merck
All Photos(1)

Documents

282839

Sigma-Aldrich

Ethyl vinyl sulfone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5S(O)2CH=CH2
CAS Number:
Molecular Weight:
120.17
Beilstein:
1745130
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.463 (lit.)

bp

118-119 °C/22 mmHg (lit.)

density

1.151 g/mL at 25 °C (lit.)

SMILES string

CCS(=O)(=O)C=C

InChI

1S/C4H8O2S/c1-3-7(5,6)4-2/h3H,1,4H2,2H3

InChI key

BJEWLOAZFAGNPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethly vinyl sulfone alkylates ε-amino groups of lysine side chains and imidazole groups of histidine residues in proteins. Chemical modification of bovine serum albumin by ethyl vinyl sulfone has been studied by X-ray photoelectron spectroscopy.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X-ray photoelectron spectroscopy of BSA and ethyl vinyl sulfone modified BSA.
M M Millard et al.
Biochemical and biophysical research communications, 70(2), 445-451 (1976-05-17)
M Friedman et al.
International journal of peptide and protein research, 7(6), 481-486 (1975-01-01)
Ethly vinyl sulfone (EVS) alkylates xi-amino groups of lysine side chains and imidazole groups of histidine residues in proteins. Amino acid analysis of hydrolyzates of EVS-treated polylysine shows that lysine forms two derivatives, presumably xi-N-(ethylsulfonylethyl)lysine and xi, xi, N,N-bis(ethylsulfonylethyl)lysine that
M S Masri et al.
Journal of protein chemistry, 7(1), 49-54 (1988-02-01)
Disulfide bonds of bovine serum albumin and wool were reduced by n-tributylphosphine to sulfhydryl groups that were then modified by methyl or ethyl vinyl sulfone in a nucleophilic addition reaction to S-(beta-ethylsulfonylmethyl)-L-cysteine and S(beta-ethylsulfonylethyl)-L-cysteine, respectively. The reductive alkylation was carried

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service