Skip to Content
Merck
All Photos(1)

Key Documents

273848

Sigma-Aldrich

2,6-Dimethyl-4-heptanone

99%

Synonym(s):

Diisobutyl ketone, Isovalerone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COCH2CH(CH3)2
CAS Number:
Molecular Weight:
142.24
Beilstein:
1743163
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.9 (vs air)

vapor pressure

1.7 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

745 °F

expl. lim.

0.8-6.2 %, 100 °F

refractive index

n20/D 1.412 (lit.)

bp

165-170 °C (lit.)

density

0.808 g/mL at 25 °C (lit.)

SMILES string

CC(C)CC(=O)CC(C)C

InChI

1S/C9H18O/c1-7(2)5-9(10)6-8(3)4/h7-8H,5-6H2,1-4H3

InChI key

PTTPXKJBFFKCEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,6-Dimethyl-4-heptanone is a volatile organic compound present in the gaseous stream imitating emissions generated at a leather industry. Its biological treatment in a suspended-growth bioreactor (SGB) has been studied.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F

Flash Point(C)

49 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M F Carvalho et al.
Chemosphere, 74(2), 232-238 (2008-11-08)
A suspended-growth bioreactor (SGB) was operated for the treatment of a gaseous stream mimicking emissions generated at a leather industrial company. The main volatile organic compounds (VOCs) present in the gaseous stream consisted of 1-methoxy-2-propanol, 2,6-dimethyl-4-heptanone, 2-butoxyethanol, toluene and butylacetate.
Marte Balchen et al.
Analytica chimica acta, 716, 16-23 (2012-01-31)
A totally new concept based on electrokinetic migration was evaluated for the extraction of three biologically active peptides from human plasma. Angiotensin 2, leu-enkephalin, and endomorphin 1 migrated from a diluted human plasma sample (2 mL, positive electrode), through a
[Thermal desorption gas chromatography for determination of diisobutyl ketone in air of working places].
Wei Chen et al.
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases, 26(2), 110-112 (2008-05-01)
R F White et al.
Environmental research, 61(1), 117-123 (1993-04-01)
The objective of this investigation was to examine cerebral magnetic resonance imaging (MRI) pathology and functional deficits demonstrated by neuropsychological testing in cases of toxic encephalopathy. Two subjects, occupationally exposed to toxic chemicals, were studied. As part of their neurological
D E Dodd et al.
Toxicology and industrial health, 3(4), 443-457 (1987-12-01)
Four groups, each consisting of 10 male and 10 female Fischer-344 rats, were exposed 6 h/day, 5 days/week, for 9 days to diisobutyl ketone (DIBK) vapor at concentrations of 905, 300, 98, or 0 (control) ppm. An additional 10 rats/sex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service