All Photos(1)
About This Item
Linear Formula:
CH3C≡CCH2CH3
CAS Number:
Molecular Weight:
68.12
Beilstein:
1696938
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor pressure
393 mmHg ( 37.7 °C)
Assay
98%
form
liquid
refractive index
n20/D 1.404 (lit.)
bp
56-57 °C (lit.)
mp
−109 °C (lit.)
density
0.71 g/mL at 25 °C (lit.)
SMILES string
CCC#CC
InChI
1S/C5H8/c1-3-5-4-2/h3H2,1-2H3
InChI key
NKTDTMONXHODTI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
2-Pentyne was used to test the catalytic activity of nanoparticles of palladium supported on bacterial biomass, bio-Pd.
accessory
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-23.8 °F - closed cup
Flash Point(C)
-31 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Palladium supported on bacterial biomass as a novel heterogeneous catalyst: A comparison of Pd/Al2O3 and bio-Pd in the hydrogenation of 2-pentyne.
Bennett JA, et al.
Chemical Engineering Science, 65(1), 282-290 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service