Skip to Content
Merck
All Photos(1)

Key Documents

266914

Sigma-Aldrich

Molybdenum

wire, diam. 1.0 mm, 99.95% trace metals basis

Synonym(s):

Molybdenum element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mo
CAS Number:
Molecular Weight:
95.94
EC Number:
MDL number:
UNSPSC Code:
12141727
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95% trace metals basis

form

wire

resistivity

5.0 μΩ-cm, 20°C

diam.

1.0 mm

bp

4612 °C (lit.)

mp

2617 °C (lit.)

density

10.3 g/mL at 25 °C (lit.)

SMILES string

[Mo]

InChI

1S/Mo

InChI key

ZOKXTWBITQBERF-UHFFFAOYSA-N

Related Categories

Quantity

40 g = 5 m

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Joyee Mitra et al.
Inorganic chemistry, 52(6), 3032-3042 (2013-03-07)
A series of [Mo(IV)O(mnt)(SR)(N-N)](-) (mnt = maleonitriledithiolate; R = Ph, nap, p-Cl-Ph, p-CO2H-Ph, and p-NO2-Ph; N-N = 2,2'-bipyridine (bipy) and 1,10-phenanthroline (phen)) complexes analogous to the reduced active site of enzymes of the sulfite oxidase family has been synthesized and
Phillip Ringel et al.
The Journal of biological chemistry, 288(20), 14657-14671 (2013-03-30)
Nitrate reductase (NR) is a complex molybdenum cofactor (Moco)-dependent homodimeric metalloenzyme that is vitally important for autotrophic organism as it catalyzes the first and rate-limiting step of nitrate assimilation. Beside Moco, eukaryotic NR also binds FAD and heme as additional
M Jake Pushie et al.
Inorganic chemistry, 52(6), 2830-2837 (2013-03-01)
Molybdenum K-edge X-ray absorption spectroscopy (XAS) has been used to probe the structure of a Mo(V) species that has been suggested to be a catalytic intermediate in the reaction of dimethyl sulfoxide (DMSO) reductase with the alternative substrate trimethylamine N-oxide
Elena Vigonsky et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(14), 5440-5445 (2013-03-21)
In all kingdoms of life, ATP Binding Cassette (ABC) transporters participate in many physiological and pathological processes. Despite the diversity of their functions, they have been considered to operate by a largely conserved mechanism. One deviant is the vitamin B12
K Cory Macleod et al.
Nature chemistry, 5(7), 559-565 (2013-06-22)
The reduction of gaseous nitrogen is a challenge for industrial, biological and synthetic chemists. Major goals include understanding the formation of ammonia for agriculture, and forming N-C and N-Si bonds for the synthesis of fine chemicals. The iron-molybdenum active site

Articles

Biomedical implants are essentially foreign substances within the human body that must survive many years’ exposure to demanding mechanical and physiological conditions. Despite these challenges, metal implants have been widely used to substitute for or rebuild hard tissues such as bones and teeth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service