266353
Zinc
pieces, 2-14 mesh, 99.9% trace metals basis
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor pressure
1 mmHg ( 487 °C)
Assay
99.9% trace metals basis
form
pieces
reaction suitability
core: zinc
reagent type: catalyst
resistivity
5.8 μΩ-cm, 20°C
particle size
2-14 mesh
bp
907 °C (lit.)
mp
420 °C (lit.)
density
7.133 g/mL at 25 °C (lit.)
SMILES string
[Zn]
InChI
1S/Zn
InChI key
HCHKCACWOHOZIP-UHFFFAOYSA-N
Application
Reducing agent; reagent used for preparation of organozinc reagents
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Knochel, P.
Chemical Reviews, 93, 217-217 (1993)
M E Kuyumcu et al.
European review for medical and pharmacological sciences, 17(11), 1467-1471 (2013-06-19)
Zinc is one of the most important elements for human body. Zinc deficiency can occur in any age, if it is seen in elderly its clinical results can be more harmful due to already diminished functions. Some studies showed zinc
Nalini Pandey et al.
Indian journal of experimental biology, 51(7), 548-555 (2013-08-01)
Black gram plants subjected to varying levels of Zn supply (0.01 to 10 microM Zn) showed optimum growth and dry matter yield in plants receiving 1 microM Zn. The dry matter yield of plants decreased in plants receiving 0.01 and
Sara Eyal et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 50(5), 798-806 (2009-05-01)
Studies in rodents indicate that the disruption of P-glycoprotein (P-gp) function increases drug distribution into the developing fetus and organs such as the brain. To simultaneously and serially evaluate the effect of P-gp activity and inhibition on the tissue distribution
W Reainthippayasakul et al.
Journal of nanoscience and nanotechnology, 13(5), 3617-3621 (2013-07-19)
Conjugated meso-alkynyl 5,15-dimesitylporphyrin metal complexes have been synthesized by Sonogashira coupling reaction in good yields. Alkynyl groups were chosen as a link at the meso positions in order to extend the pi-conjugated length of porphyrin rings. These synthesized porphyrin derivatives
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service