Skip to Content
Merck
All Photos(2)

Key Documents

263354

Sigma-Aldrich

2-Bromo-5-methylpyridine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6BrN
CAS Number:
Molecular Weight:
172.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

bp

95-96 °C/12.5 mmHg (lit.)

mp

41-43 °C (lit.)

functional group

bromo

SMILES string

Cc1ccc(Br)nc1

InChI

1S/C6H6BrN/c1-5-2-3-6(7)8-4-5/h2-4H,1H3

InChI key

YWNJQQNBJQUKME-UHFFFAOYSA-N

Application

2-Bromo-5-methylpyridine has been used as starting reagent for the synthesis of:
  • 5,5′-dimethyl-2,2′-bipyridine
  • 5-methyl-2,2′:6′,2″-terpyridine
  • 5-bromo-5″-methyl-2,2′:6′,2″-terpyridine

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of two covalently linked bis (2, 2': 6', 2''-terpyridine)(terpy) chelating ligands with different length spacers, comparison of the crystal structures of their mononuclear nickel (II) complexes, and kinetic and mechanistic studies of the reaction of one ligand with [Fe (terpy) 2] 2+.
Priimov GU, et al.
J. Chem. Soc., Dalton Trans., 4, 445-449 (2000)
High yield synthesis of 5, 5'-dimethyl-2, 2'-bipyridine and 5, 5 ?-dimethyl-2, 2': 6', 2 ?-terpyridine and some bisfunctionalization reactions using N-bromosuccinimide.
Schubert US, et al.
Synthesis, 1999(5), 779-782 (1999)
Synthesis of a novel ditopic ligand incorporating directly bonded 1, 10-phenanthroline and 2, 2': 6', 2 ?-terpyridine units.
Gavi?a P and Tatay S.
Tetrahedron Letters, 47(20), 3471-3473 (2006)
Ben Yiu-Wing Wong et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 22(42), 15095-15106 (2016-09-07)
A series of air-stable spiro-fused ladder-type boron(III) compounds has been designed, synthesized, and the electrochemistry and photophysical behavior have been characterized. By simply varying the substituents on the pyridine ring and extending the π-conjugation of the spiro framework, the emission

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service