Skip to Content
Merck
All Photos(1)

Key Documents

260932

Sigma-Aldrich

IR-140

Dye content 95 %

Synonym(s):

5,5′-Dichloro-11-diphenylamino-3,3′-diethyl-10,12-ethylenethiatricarbocyanine perchlorate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H34Cl3N3O4S2
CAS Number:
Molecular Weight:
779.19
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

autoignition temp.

1075 °F

composition

Dye content, 95%

mp

250-252 °C (lit.)

λmax

823 nm

SMILES string

[O-]Cl(=O)(=O)=O.CCN1C(/Sc2ccc(Cl)cc12)=C\C=C3/CCC(\C=C\c4sc5ccc(Cl)cc5[n+]4CC)=C3N(c6ccccc6)c7ccccc7

InChI

1S/C39H34Cl2N3S2.ClHO4/c1-3-42-33-25-29(40)19-21-35(33)45-37(42)23-17-27-15-16-28(18-24-38-43(4-2)34-26-30(41)20-22-36(34)46-38)39(27)44(31-11-7-5-8-12-31)32-13-9-6-10-14-32;2-1(3,4)5/h5-14,17-26H,3-4,15-16H2,1-2H3;(H,2,3,4,5)/q+1;/p-1

InChI key

UVLDECUUBLLYRG-UHFFFAOYSA-M

Application

Suitable as laser dye

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yogita Silori et al.
The journal of physical chemistry. B, 124(31), 6825-6834 (2020-07-10)
Photophysical properties of tricarbocyanine dyes in various solvents have been widely investigated using a variety of spectroscopic tools. However, the presence of several ground-state isomers and interconversion between these isomers on an ultrafast timescale upon photoexcitation render unambiguous assignment of
Ankun Yang et al.
Nature communications, 6, 6939-6939 (2015-04-22)
Plasmon lasers can support ultrasmall mode confinement and ultrafast dynamics with device feature sizes below the diffraction limit. However, most plasmon-based nanolasers rely on solid gain materials (inorganic semiconducting nanowire or organic dye in a solid matrix) that preclude the
Ken-Ichi Yamashita et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(16), 3633-3640 (2019-12-28)
Facile redox-induced aromatic-antiaromatic interconversions were accomplished by using β-tetracyano-21,23-dithiaporphyrin (CN4 S2 Por). Introduced cyano groups not only increased the reduction potential of the porphyrin core but also stabilized the antiaromatic isophlorin (CN4 S2 Iph) by π conjugation. The reduction of

Articles

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service