Skip to Content
Merck
All Photos(1)

Key Documents

258555

Sigma-Aldrich

p-Phenylene diisothiocyanate

98%

Synonym(s):

1,4-Diisothiocyanatobenzene, Bitoscanate, PDITC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(NCS)2
CAS Number:
Molecular Weight:
192.26
Beilstein:
2414340
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

reagent type: cross-linking reagent

mp

129-131 °C (lit.)

SMILES string

S=C=Nc1ccc(cc1)N=C=S

InChI

1S/C8H4N2S2/c11-5-9-7-1-2-8(4-3-7)10-6-12/h1-4H

InChI key

OMWQUXGVXQELIX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

p-Phenylene diisothiocyanate can be used in various applications including the following:
  • Synthesis of hydroxamate containing chelators for labeling antibodies with 89Zr and 68Ga for imaging with positron emission tomography (immunoPET).
  • Synthesis of fluorescent chemosensors for Hg2+ sensing in living cells.
  • Synthesis of polythiourea derivatives as capacitor dielectric materials.
  • Synthesis of 4-isothiocyanato-4′-nitrodiphenylamine, an anthelmintic agent, popularly known as amoscanate.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multifunctional Desferrichrome Analogues as Versatile 89Zr (IV) Chelators for ImmunoPET Probe Development.
Adams CJ et al.
Molecular Pharmaceutics, 14(8), 2831-2842 (2017)
Design, synthesis and evaluation of novel bifunctional tetrahydroxamate chelators for PET imaging of 89Zr-labeled antibodies.
Rousseau J, et al.
Bioorganic & Medicinal Chemistry, 27(4), 708-712 (2018)
A New Synthesis Of 4-Isothiocyanato-4'-Nitrodiphenylamine (Amoscanate).
Viswanathan N and Desai RC.
Indian J. Chem. B, 20(4), 308-310 (1981)
Chengwei Li et al.
Nanoscale research letters, 15(1), 36-36 (2020-02-08)
Construct dielectric films with high energy density and efficiency are the key factor to fabricate high-performance dielectric film capacitors. In this paper, an all organic composite film was constructed based on high dielectric polymer and linear dielectric polymer. After the
Rational design and synthesis of polythioureas as capacitor dielectrics.
Ma R, et al.
Journal of Material Chemistry A, 3(28), 14845-14852 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service