Skip to Content
Merck

Skip To

257575

2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine manganese(III) chloride

97%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C36H44ClMnN4
CAS Number:
Molecular Weight:
623.15
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12352103
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

97%

form

powder

reaction suitability

reagent type: catalyst
core: manganese

λmax

459 nm

SMILES string

CCc1c(CC)c2cc3c(CC)c(CC)c4cc5nc(cc6c(CC)c(CC)c(cc1n2)n6[Mn](Cl)n34)c(CC)c5CC

InChI

1S/C36H44N4.ClH.Mn/c1-9-21-22(10-2)30-18-32-25(13-5)26(14-6)34(39-32)20-36-28(16-8)27(15-7)35(40-36)19-33-24(12-4)23(11-3)31(38-33)17-29(21)37-30;;/h17-20H,9-16H2,1-8H3;1H;/q-2;;+3/p-1/b29-17-,30-18-,31-17-,32-18-,33-19-,34-20-,35-19-,36-20-;;

InChI key

ZMCDJCAIDCCCKY-YAJYDHHFSA-M

General description

2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine manganese(III) chloride is formed by metalating 2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine with manganese (III) chloride.1

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
258423269948392014
description

97%

description

97%

description

97%

description

Dye content 85 %

form

powder

form

-

form

solid

form

solid

assay

97%

assay

97%

assay

97%

assay

-

reaction suitability

reagent type: catalyst
core: manganese

reaction suitability

-

reaction suitability

reagent type: catalyst
core: nickel

reaction suitability

reagent type: catalyst
core: palladium

λmax

459 nm

λmax

400 nm

λmax

392 nm

λmax

393 nm


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters