All Photos(1)
About This Item
Linear Formula:
C6H5C6H4Br
CAS Number:
Molecular Weight:
233.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
liquid
refractive index
n20/D 1.639 (lit.)
bp
103-104 °C/0.2 mmHg (lit.)
density
1.398 g/mL at 25 °C (lit.)
functional group
bromo
phenyl
SMILES string
Brc1cccc(c1)-c2ccccc2
InChI
1S/C12H9Br/c13-12-8-4-7-11(9-12)10-5-2-1-3-6-10/h1-9H
InChI key
USYQKCQEVBFJRP-UHFFFAOYSA-N
General description
Electrochemical immunosensor based on polydopamine coated prussian blue-mesoporous carbon nanocomposite for rapid screening of 3-bromobiphenyl has been reported.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Zihong Sun et al.
Biosensors & bioelectronics, 59, 99-105 (2014-04-09)
A sensitive electrochemical immunosensor for 3-bromobiphenyl (3-BBP) detection was constructed by employing a new polydopamine coated prussian blue-mesoporous carbon (PDOP/PB/CMK-3) nanocomposite as the substrate platform and multi-horseradish peroxidase-double helix carbon nanotubes-secondary antibody (multi-HRP-DHCNTs-Ab2) as the signal label. PB/CMK-3 was firstly
Asami Yoshii et al.
Chemistry, an Asian journal, 15(14), 2181-2186 (2020-05-26)
Oligo-meta-phenylenes have been designed and synthesized as multipotent base materials of single-layer organic light-emitting devices. Simple molecular structures of oligo-meta-phenylenes composed of linear phenylene arrays benefited from the wealth of modern reactions available for biaryl couplings and were concisely synthesized
Man Wang et al.
Journal of chromatography. A, 1412, 12-21 (2015-08-19)
A new magnetic porous carbon-doped graphitic carbon nitride nanocomposite and experimental strategies were environment-friendly designed for solid phase extraction of brominated flame retardants from water sample. The easily synthesized and low cost nanocomposite was characterized using techniques, including Fourier transform
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service