Skip to Content
Merck
All Photos(3)

Key Documents

254061

Sigma-Aldrich

Europium(III) nitrate hydrate

99.99% trace metals basis

Synonym(s):

Europic nitrate hydrate, Europium trinitrate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Eu(NO3)3 · xH2O
CAS Number:
Molecular Weight:
337.98 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
form:
solid

Quality Level

Assay

99.99% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: europium

impurities

≤150.0 ppm Trace Rare Earth Analysis

SMILES string

[Eu+3].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Eu.3NO3.5H2O/c;3*2-1(3)4;;;;;/h;;;;5*1H2/q+3;3*-1;;;;;

InChI key

BBGDGFQCQRFYCP-UHFFFAOYSA-N

Application

  • A novel strategy for the synthesis of samarium/europium-metal organic frameworks: This study explores the utilization of Samarium and Europium nitrate hydrates in the development of metal-organic frameworks for luminescent sensing. These frameworks are employed for detecting substances like acetone, Cr3+, and Pb2+ in real samples, showcasing potential applications in environmental monitoring and safety (Syed, 2024).

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lifu Xiao et al.
Analytical methods : advancing methods and applications, 10(1), 22-29 (2018-02-17)
G-protein-coupled receptor 120 (GPR120), as a member of the rhodopsin family of G-protein-coupled receptors, has been shown to function as a sensor for dietary fat in the gustatory and digestive systems. Its specific role in the chemoreception of fatty acids
Bhavesh D Kevadiya et al.
Theranostics, 8(1), 256-276 (2018-01-02)
Long-acting slow effective release antiretroviral therapy (LASER ART) was developed to improve patient regimen adherence, prevent new infections, and facilitate drug delivery to human immunodeficiency virus cell and tissue reservoirs. In an effort to facilitate LASER ART development, "multimodal imaging
Yujuan He et al.
Nanotechnology, 31(23), 235603-235603 (2020-02-14)
Yttrium orthovanadate (YVO4) has been widely used as a host material for low- and medium-power diode-pumped solid-state lasers due to its excellent thermal, mechanical, and optical properties. This work demonstrates the synthesis and site symmetry tunning of polycrystalline YVO4:Eu nanoparticles
Vishnu Sravan Bollu et al.
Nanotoxicology, 10(4), 413-425 (2015-10-08)
Our group already demonstrated that europium hydroxide nanorods (EHNs) show none or mild toxicity in C57BL/6 mice even at high dose and exhibited excellent pro-angiogenic activity towards in vitro and in vivo models. In the present study, we evaluated the
Bhavesh D Kevadiya et al.
Acta biomaterialia, 49, 507-520 (2016-12-06)
The size, shape and chemical composition of europium (Eu A novel europium (Eu

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service