Skip to Content
Merck
All Photos(1)

Documents

250619

Sigma-Aldrich

4-Amino-1-naphthalenesulfonic acid

97%

Synonym(s):

1-Naphthylamine-4-sulfonic acid, Naphthionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC10H6SO3H
CAS Number:
Molecular Weight:
223.25
Beilstein:
1971299
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

≥300 °C (lit.)

solubility

water: very slightly soluble(lit.)

SMILES string

Nc1ccc(c2ccccc12)S(O)(=O)=O

InChI

1S/C10H9NO3S/c11-9-5-6-10(15(12,13)14)8-4-2-1-3-7(8)9/h1-6H,11H2,(H,12,13,14)

InChI key

NRZRRZAVMCAKEP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Amino-1-naphthalenesulfonic acid is a diazo compound. It is also known as Piria′s acid.

Application

4-Amino-1-naphthalenesulfonic acid has potent therapeutic applications. Its sodium salt has been used as non-toxic hemostatic.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tracing polar benzene-and naphthalenesulfonates in untreated industrial effluents and water treatment works by ion-pair chromatography-fluorescence and electrospray-mass spectrometry.
Alonso MC and Barcelo D.
Analytica Chimica Acta, 400(1), 211-231 (1999)
Circulating naphthionic acid in nonpregnant and pregnant rats after feeding amaranth.
R F Willes et al.
Toxicology and applied pharmacology, 54(2), 276-284 (1980-06-30)
C Juárez-Ramírez et al.
Journal of industrial microbiology & biotechnology, 39(8), 1169-1177 (2012-04-12)
By decolorization of azo dyes, caused by reductive cleavage of the azo linkage, toxic or recalcitrant amines are generated. The present study deals with the effect of the inflowing medium composition (C:N ratio) on the kinetic behavior of a bacterial
A S Rodrigues et al.
Chemosphere, 205, 433-442 (2018-05-01)
The electrodegradation of the 4-aminonaphthalene-1-sulfonic acid (4AN1S), 5-aminonaphthalene-2-sulfonic acid (5AN2S) and 8-aminonaphthalene-2-sulfonic acid (8AN2S) was studied, using two electrode materials as anode, BDD and Ti/Pt/PbO2, and two different electrolytes, sodium sulfate and sodium chloride. The highest COD removal rates were
Yinggang Luo et al.
The Journal of biological chemistry, 283(21), 14694-14702 (2008-04-05)
Neocarzinostatin, a clinical anticancer drug, is the archetypal member of the chromoprotein family of enediyne antitumor antibiotics that are composed of a nonprotein chromophore and an apoprotein. The neocarzinostatin chromophore consists of a nine-membered enediyne core, a deoxyaminosugar, and a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service