Skip to Content
Merck
All Photos(1)

Key Documents

250023

Sigma-Aldrich

(S)-(+)-1-Bromo-2-methylbutane

99%

Synonym(s):

(S)-(+)-2-Methylbutyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)CH2Br
CAS Number:
Molecular Weight:
151.04
Beilstein:
1718815
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

optical activity

[α]21/D +4.5°, c = 5 in chloroform

refractive index

n20/D 1.445 (lit.)

bp

121-122 °C (lit.)

density

1.223 g/mL at 25 °C (lit.)

functional group

alkyl halide
bromo

SMILES string

CC[C@H](C)CBr

InChI

1S/C5H11Br/c1-3-5(2)4-6/h5H,3-4H2,1-2H3/t5-/m0/s1

InChI key

XKVLZBNEPALHIO-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For the synthesis of chiral nematic liquid crystals and optically active Grignard reagents.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nathan T Derstine et al.
Journal of insect science (Online), 12, 93-93 (2013-02-21)
Two detection methods for the little fire ant, Wasmannia auropunctata (Roger) (Hymenoptera: Formicidae), both employing the pheromone attractant 2,5-dimethyl-3-(2-methylbutyl)pyrazine (2-MeBu-diMePy), were compared with peanut butter based detection, in order to evaluate differences in species specificity and detection reliability. Trapping was
Journal of Materials Chemistry, 3, 609-609 (1993)
Zh. Org. Khim., 28, 1173-1173 (1992)
Journal of Materials Chemistry, 2, 119-119 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service