Skip to Content
Merck
All Photos(2)

Key Documents

249319

Sigma-Aldrich

4-Amino-3-nitrophenol

98%

Synonym(s):

4-Hydroxy-2-nitroaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(NO2)OH
CAS Number:
Molecular Weight:
154.12
Beilstein:
2210196
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

mp

150-154 °C (lit.)

functional group

nitro

SMILES string

Nc1ccc(O)cc1[N+]([O-])=O

InChI

1S/C6H6N2O3/c7-5-2-1-4(9)3-6(5)8(10)11/h1-3,9H,7H2

InChI key

IQXUIDYRTHQTET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Amino-3-nitrophenol was used in the synthesis of benzimidazole based Factor Xa (trypsin-like serine protease) inhibitors[1] and 4-(4-amino-3-nitrophenoxy)phthalonitrile[2]. It was also employed as matrix during visible matrix-assisted laser desorption/ionization mass spectrometry[3].

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization, and electrical, electrochemical and gas sensing properties of a novel cyclic borazine derivative containing three phthalocyaninato zinc (II) macrocycles.
Ozer M, et al.
Synt. Metals, 155(1), 222-231 (2005)
Lorianne R Shultz et al.
Molecules (Basel, Switzerland), 25(1) (2019-12-29)
Para-, or 4-nitrophenol, and related nitroaromatics are broadly used compounds in industrial processes and as a result are among the most common anthropogenic pollutants in aqueous industrial effluent; this requires development of practical remediation strategies. Their catalytic reduction to the
Z Zhao et al.
Bioorganic & medicinal chemistry letters, 10(9), 963-966 (2000-06-15)
Inhibitors based on the benzimidazole scaffold showed subnanomolar potency against Factor Xa with 500-1000-fold selectivity against thrombin and 50-100-fold selectivity against trypsin. The 2-substituent on the benzimidazole ring had a strong impact on the FXa inhibitory activity. Crystallography studies suggest
Qiaoran Liu et al.
Journal of colloid and interface science, 569, 12-21 (2020-02-26)
Organic contaminants, dyes and antibiotics, discharged in wastewater systems, have posed great threats to the sustainability of the ecosystem. This study was performed to prepare graphitic carbon nitride (GCN) nanocomposites modified by nanocarbons, including carbon quantum dots (CQD), carbon nanotube
Lee Chuin Chen et al.
Rapid communications in mass spectrometry : RCM, 21(24), 4129-4134 (2007-11-21)
Visible matrix-assisted laser desorption/ionization (VIS-MALDI) was performed using 2-amino-3-nitrophenol as matrix. The matrix is of near-neutral pH, and has an optical absorption band in the near-UV and visible region. A frequency-doubled Nd:YAG laser operated at 532 nm wavelength was used

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service