Skip to Content
Merck
All Photos(3)

Documents

245267

Sigma-Aldrich

Manganese(0) carbonyl

98%

Synonym(s):

Decacarbonyl dimanganese(0), Dimanganese(0) decacarbonyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mn2(CO)10
CAS Number:
Molecular Weight:
389.98
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

powder or crystals

reaction suitability

core: manganese
reagent type: catalyst

mp

154 °C (lit.)

storage temp.

2-8°C

SMILES string

[Mn].[Mn].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

InChI

1S/10CO.2Mn/c10*1-2;;

InChI key

QFEOTYVTTQCYAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Manganese(0) carbonyl, also known as Decacarbonyl dimanganese exhibits interesting electrochemicalproperties owing to the presence of carbonyl ligands and the manganese metalcenters. In electrochemistry, this compound can act as a carbonyl source andpotentially participate in redox reactions.

Application

Manganese(0) carbonyl can be used:
  • As a precursor to prepare Ni2–xMnxP nanoparticles to fabricate a catalyst for electrochemical oxygen evolution reaction(OER).
  • To fabricate an ultra-sensitive manganese doped MoS2 electrochemical dopamine sensor via modified chemical vapor deposition(CVD) method.
  • As a reducing agent to synthesize Pt-Co electrocatalyst for oxygen reduction reactions in fuel cells. Mn and CO atoms produced during decomposition acts as reducing and surface structure guiding agents respectively.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ni2?xMnxP Nanoparticles as Earth-Abundant Precatalysts for Electrochemical Water Oxidation
Fatemeh Aghabozorgi, et al
Chemistry of Materials, 36, 2461-2472 (2024)
Doping and decorating 2D materials for biosensing: benefits and drawbacks
Hui Ling Chia, et al.
Advanced Functional Materials , 31, 2102555-2102555 (2021)
Pt-Co Electrocatalysts: Syntheses, Morphologies, and Applications
Hao Chen, et al.
Small, 18, 2204100-2204100 (2022)

Articles

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service