Skip to Content
Merck
All Photos(2)

Key Documents

241032

Sigma-Aldrich

Bromoform

contains 60-120 ppm 2-methyl-2-butene as stabilizer, 99%

Synonym(s):

Tribromomethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHBr3
CAS Number:
Molecular Weight:
252.73
Beilstein:
1731048
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

8.7 (vs air)

vapor pressure

5 mmHg ( 20 °C)

Assay

99%

form

liquid

contains

60-120 ppm 2-methyl-2-butene as stabilizer

refractive index

n20/D 1.595 (lit.)

bp

146-150 °C (lit.)

mp

5-8 °C (lit.)

solubility

water: soluble 800 part(lit.)
acetone: miscible(lit.)
alcohol: miscible(lit.)
benzene: miscible(lit.)
chloroform: miscible(lit.)
diethyl ether: miscible(lit.)
oil: miscible(lit.)
petroleum ether: miscible(lit.)

density

2.89 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

BrC(Br)Br

InChI

1S/CHBr3/c2-1(3)4/h1H

InChI key

DIKBFYAXUHHXCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Bromoform (CHBr3), also known as Tribromomethane, is a very short-lived substance (VSLS) and an important precursor of reactive bromine species (BrOx). It is widely used as a laboratory reagent and as an industrial extraction solvent. It can also be used as a sedative and to create block copolymers.

Application

Bromoform (Tribromomethane) was used in the synthesis of α-tribromomethylated N-sulfonylamines.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

106.3 °F - closed cup - (own results)

Flash Point(C)

41.3 °C - closed cup - (own results)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elumalai Gopi et al.
Organic & biomolecular chemistry, 12(17), 2769-2777 (2014-03-29)
Mg-mediated addition of bromoform to electron deficient imines such as N-sulfonylimines affords α-tribromomethylated N-sulfonylamines in good to excellent yields. The procedure could be further simplified by transforming the imine precursors, α-sulfonyl-N-tosyl- and Boc-amines, in one pot to the corresponding α-tribromomethyl
Bichismita Sahu et al.
The Journal of organic chemistry, 74(6), 2601-2604 (2009-02-26)
Addition of bromoform to conjugated nitroalkenes in the presence of Mg provided beta-tribromomethyl nitroalkanes in good to excellent yields and diastereoselectivity. These novel Michael adducts, formed under radical conditions, underwent elimination of HBr in the same pot under reflux to
Lisa George et al.
The Journal of chemical physics, 135(12), 124503-124503 (2011-10-07)
Iso-polyhalomethanes are known reactive intermediates that play a pivotal role in the photochemistry of halomethanes in condensed phases. In this work, iso-bromoform (iso-CHBr(3)) and its deuterated isotopomer were characterized by matrix isolation infrared and UV/visible spectroscopy, supported by ab initio
M Valcke et al.
Regulatory toxicology and pharmacology : RTP, 59(2), 258-269 (2010-10-26)
The objective of this study was to assess the impact of the exposure route on the human kinetic adjustment factor (HKAF), for which a default value of 3.16 is used in non-cancer risk assessment. A multi-route PBPK model was modified
Jong-Ryul Park et al.
Biointerphases, 16(1), 011001-011001 (2021-01-07)
Poly(2-alkyl-2-oxazoline) (PAOx) hydrogels are tailorable synthetic materials with demonstrated biomedical applications, thanks to their excellent biocompatibility and tunable properties. However, their use as injectable hydrogels is challenging as it requires invasive surgical procedures to insert the formed hydrogel into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service