Skip to Content
Merck
All Photos(3)

Key Documents

232211

Sigma-Aldrich

4-Pentynoic acid

95%

Synonym(s):

Propargylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH≡CCH2CH2COOH
CAS Number:
Molecular Weight:
98.10
Beilstein:
1742047
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

bp

110 °C/30 mmHg (lit.)

mp

54-57 °C (lit.)

functional group

carboxylic acid

storage temp.

2-8°C

SMILES string

OC(=O)CCC#C

InChI

1S/C5H6O2/c1-2-3-4-5(6)7/h1H,3-4H2,(H,6,7)

InChI key

MLBYLEUJXUBIJJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Pentynoic acid undergoes copper-catalyzed intramolecular cyclizations to form enol lactones. It also reacts with 1-bromo-1-alkynes in the presence of a Pd catalyst to yield biologically active ynenol lactones.

Application

4-Pentynoic acid was used:
  • as building block for the synthesis of library of eight sequence-defined model oligomers
  • in one-pot synthesis of the complex polycyclic heterocycles benzo[4,5]imidazo[1,2-c]pyrrolo[1,2-a]quinazolinone derivatives
  • in the synthesis of various allenenols lactones [5(E)-(2-allenylidene)-tetrahydro-2-furanones]
  • in the synthesis of a cyctotoxic macrolide by ring-closing metathesis of a bis acetylene

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-catalyzed synthesis of new unsaturated exo-enol lactones with potential biological activity.
Bouyssi D, et al.
Tetrahedron Letters, 34(19), 3129-3130 (1993)
Thomas L Mindt et al.
The Journal of organic chemistry, 72(26), 10247-10250 (2007-11-30)
Alkynoic acids, in particular, 4-pentynoic acid derivatives, undergo intramolecular cyclizations to enol lactones under reaction conditions typically applied for the Cu(I)-catalyzed cycloaddition of terminal alkynes and azides (click chemistry). Starting from appropriate alkynoic acid derivatives, either enol lactones or 1,2,3-triazole
Tetrahedron Letters, 33, 2811-2811 (1992)
Xin Zhou et al.
Acta biomaterialia, 54, 128-137 (2017-03-13)
Wound healing dressings are increasingly needed clinically due to the large number of skin damage annually. Nitric oxide (NO) plays a key role in promoting wound healing, thus biomaterials with NO-releasing property receive increasing attention as ideal wound dressing. In
Thanh Tam Trinh et al.
Macromolecular rapid communications, 35(2), 141-145 (2013-12-18)
A library of eight sequence-defined model oligomers, whose sequence is based on a (0,1) binary code, is prepared through chemoselective repeating cycles of amidification and copper-assisted alkyne-azide cycloaddition reactions from a non-modified Wang resin. This library is constructed from two

Articles

Click chemistry, and the copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) in particular, is a powerful new synthetic tool in polymer chemistry and material science.

Click chemistry, and the copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) in particular, is a powerful new synthetic tool in polymer chemistry and material science.

Click chemistry, and the copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) in particular, is a powerful new synthetic tool in polymer chemistry and material science.

Click chemistry, and the copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) in particular, is a powerful new synthetic tool in polymer chemistry and material science.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service