Skip to Content
Merck
All Photos(1)

Documents

230537

Sigma-Aldrich

2-Amino-6-methylbenzoic acid

99%

Synonym(s):

6-Methylanthranilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(CH3)CO2H
CAS Number:
Molecular Weight:
151.16
Beilstein:
2803391
MDL number:
UNSPSC Code:
12352106
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

reaction suitability

reaction type: solution phase peptide synthesis

mp

128-130 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

Cc1cccc(N)c1C(O)=O

InChI

1S/C8H9NO2/c1-5-3-2-4-6(9)7(5)8(10)11/h2-4H,9H2,1H3,(H,10,11)

InChI key

XHYVBIXKORFHFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Van Haandel et al.
Journal of agricultural and food chemistry, 48(5), 1949-1954 (2000-05-23)
The potential of different peroxidase preparations for the N-demethylation of methyl N-methylanthranilate to produce the food flavor methylanthranilate (MA) was investigated. All tested peroxidase preparations were able to catalyze the N-dealkylation. The tested soybean preparations vary widely with respect to
A B Rose et al.
Genetics, 145(1), 197-205 (1997-01-01)
Nine blue fluorescent mutants of the flowering plant Arabidopsis thaliana were isolated by genetic selections and fluorescence screens. Each was shown to contain a recessive allele of trp1, a previously described locus that encodes the tryptophan biosynthetic enzyme phosphoribosylanthranilate transferase
J Li et al.
Plant physiology, 110(1), 51-59 (1996-01-01)
The first step of tryptophan biosynthesis is catalyzed by anthranilate synthase (AS), which is normally subject to feedback inhibition by tryptophan. Three independent trp5 mutants defective in the Arabidopsis thaliana AS alpha subunit structural gene ASA1 were identified by selection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service