Skip to Content
Merck
All Photos(3)

Documents

229547

Sigma-Aldrich

Ammonium cerium(IV) nitrate

≥99.99% trace metals basis

Synonym(s):

Ceric ammonium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce(NH4)2(NO3)6
CAS Number:
Molecular Weight:
548.22
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.99% trace metals basis

form

crystalline

reaction suitability

reagent type: oxidant

impurities

≤100.0 ppm Trace Metal Analysis

SMILES string

N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Ce.2HNO3.4NO3.2H3N/c;6*2-1(3)4;;/h;2*(H,2,3,4);;;;;2*1H3/q+4;;;4*-1;;

InChI key

WIBGOERAEYJBOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium cerium(IV) nitrate was used to initiate the graft polymerization of polyacrylic acid with magnetic cation exchange adsorbents, which was used in conjugation with micellar aqueous two-phase systems for protein separation.
Offers a novel route to tartronic acid derivatives via facile oxygenation of malonates.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Skin Corr. 1B - Skin Sens. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein separation with magnetic adsorbents in micellar aqueous two-phase systems.
Becker JS, et al.
Separation and Purification Technology, 65.1, 46-53 (2009)
Nair, V. et al.
Tetrahedron Letters, 39, 2801-2801 (1998)
Naoki Surugaya et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 24(3), 377-380 (2008-03-12)
A simple and rapid spectrophotometric method has been developed for the determination of Pu in highly radioactive liquid waste. This method uses Nd(III) as an internal standard, which enables us to determine the concentration of Pu and to authenticate the
Alexander M Jacobine et al.
The Journal of organic chemistry, 73(18), 7409-7412 (2008-08-13)
The generation of substituted gamma-lactones can be accomplished through application of a tandem chain extension-aldol reaction, followed by CAN-mediated oxidative cleavage of the aldol product. The oxidative cleavage requires the intermediacy of a hemiketal and the presence of an alpha-heteroatom.
Douglas B Grotjahn et al.
Journal of the American Chemical Society, 133(47), 19024-19027 (2011-11-09)
Organometallic iridium complexes have been reported as water oxidation catalysts (WOCs) in the presence of ceric ammonium nitrate (CAN). One challenge for all WOCs regardless of the metal used is stability. Here we provide evidence for extensive modification of many

Articles

Oxidation and reduction reactions are some of the most common transformations encountered in organic synthesis, and are some of the organic chemist’s most powerful tools for creating novel products. Below is a list of the most commonly used oxidizing and reducing agents currently available in our catalog.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service