227412
N,N,2,2-Tetramethyl-1,3-propanediamine
97%
Synonym(s):
N,N-Dimethylneopentanediamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.4355 (lit.)
bp
155-158 °C (lit.)
density
0.818 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
CN(C)CC(C)(C)CN
InChI
1S/C7H18N2/c1-7(2,5-8)6-9(3)4/h5-6,8H2,1-4H3
InChI key
ULDIVZQLPBUHAG-UHFFFAOYSA-N
Gene Information
rat ... Grin2a(24409)
Related Categories
General description
The pK1 and pK2 values of N,N,2,2-tetramethyl-1,3-propanediamine was determined.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
96.8 °F - closed cup
Flash Point(C)
36 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of Organic Chemistry, 40(12), 1795-1800 (1975)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service