Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CH3OC6H9(=O)
CAS Number:
Molecular Weight:
128.17
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
EC Number:
231-071-8
MDL number:
Assay:
97%
assay
97%
refractive index
n
bp
185 °C/750 mmHg (lit.)
density
1.02 g/mL at 25 °C (lit.)
functional group
ether, ketone
SMILES string
COC1CCCCC1=O
InChI
1S/C7H12O2/c1-9-7-5-3-2-4-6(7)8/h7H,2-5H2,1H3
InChI key
JYJURPHZXCLFDX-UHFFFAOYSA-N
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
156.2 °F - closed cup
flash_point_c
69 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.