Skip to Content
Merck
All Photos(1)

Key Documents

226084

Sigma-Aldrich

(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate

97%, for peptide synthesis

Synonym(s):

BOP, BOP Reagent, Castros reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22F6N6OP2
CAS Number:
Molecular Weight:
442.28
Beilstein:
4287025
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22

Product Name

(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate, 97%

Assay

97%

form

powder

reaction suitability

reaction type: Coupling Reactions

mp

>130 °C (dec.) (lit.)

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

SMILES string

F[P-](F)(F)(F)(F)F.CN(C)[P+](On1nnc2ccccc12)(N(C)C)N(C)C

InChI

1S/C12H22N6OP.F6P/c1-15(2)20(16(3)4,17(5)6)19-18-12-10-8-7-9-11(12)13-14-18;1-7(2,3,4,5)6/h7-10H,1-6H3;/q+1;-1

InChI key

MGEVGECQZUIPSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for:
Peptide coupling
Synthesis of esters
Esterification of carboxylic acids
Plasmid CAN-encapsulating liposomes
Synthesis of magnolamide for antioxidative activity

Catalyst for preparation of 9-acridinecaroboxamide derivative

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Umut Balli et al.
Journal of oral science, 58(4), 465-473 (2016-12-28)
The aim of this clinical study was to determine levels of visceral adipose tissue-derived serpin (vaspin), omentin-1, and tumor necrosis factor-alpha (TNF-α) in the gingival crevicular fluid (GCF) of obese and non-obese periodontitis patients following nonsurgical periodontal therapy. Seventy-six subjects
Brissi Zagadou et al.
Hearing research, 342, 23-38 (2016-03-13)
The current impulse noise criteria for the protection against impulse noise injury do not incorporate an objective measure of hearing protection. A new biomechanically-based model has been developed based on improvement of the Auditory Hazard Assessment Algorithm for the Human
Usmah Kawoos et al.
PloS one, 11(12), e0167510-e0167510 (2016-12-03)
Exposure to blast overpressure (BOP) activates a cascade of pathological processes including changes in intracranial pressure (ICP) and blood-brain barrier (BBB) permeability resulting in traumatic brain injury (TBI). In this study the effect of single and multiple exposures at two
Fawad Javed et al.
Photodiagnosis and photodynamic therapy, 16, 85-89 (2016-09-08)
The aim was to assess the efficacy of mechanical debridement (MD) with and without adjunct antimicrobial photodynamic therapy (aPDT) in reducing periimplant inflammation among cigarette-smokers and non-smokers. Cigarette-smokers and non-smokers were randomly divided into 2 groups. In the test-group, participants
Zhiyun Du et al.
Chemical communications (Cambridge, England), 47(46), 12488-12490 (2011-10-22)
We report here, for the first time, the BOP-mediated one-pot macrocyclization that is facilitated and guided by internally placed intramolecular H-bonds to allow for the highly selective formation of five-residue cation-binding macrocycles.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service