Skip to Content
Merck
All Photos(1)

Documents

223115

Sigma-Aldrich

Dimethyl sebacate

99%

Synonym(s):

Decanedioic acid 1,10-dimethyl ester, Dimethyl decanedioate, NSC 9415, Sebacic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCO(CH2)8COOCH3
CAS Number:
Molecular Weight:
230.30
Beilstein:
1785523
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

bp

158 °C/10 mmHg (lit.)

mp

29-31 °C (lit.)

density

0.988 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CCCCCCCCC(=O)OC

InChI

1S/C12H22O4/c1-15-11(13)9-7-5-3-4-6-8-10-12(14)16-2/h3-10H2,1-2H3

InChI key

ALOUNLDAKADEEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl sebacate (DMS) is a plasticizer that can be synthesized by esterification of sebacic acid.

Application

DMS can be used in the synthesis of decanedihydrazide which can be used as a precursor for isocyanates for the synthesis of thermoplastic polyurethanes. Transesterification of DMS with 2-ethylhexanol can be used to synthesize bis(2-ethylhexyl)sebacate for further use in petroleum based lubricants.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Novel fatty acid based di-isocyanates towards the synthesis of thermoplastic polyurethanes.
More AS, et al.
European Polymer Journal, 49(4), 823-833 (2013)
Esterification of Sebacic Acid in Near-Critical and Supercritical Methanol.
Narayan RC and Madras G
Industrial & Engineering Chemistry Research, 56(10), 2641-2649 (2017)
Experimental determination and theoretical correlation for the solubilities of dicarboxylic acid esters in supercritical carbon dioxide.
Narayan RC, et al.
Journal of Supercritical Fluids, 101, 87-94 (2015)
[Determination of dimethylacrylic acid and its ethyl ester in the air of a work area].
N Ia Smoliar
Gigiena i sanitariia, (1)(1), 51-52 (1984-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service