Skip to Content
Merck
All Photos(1)

Key Documents

222518

Sigma-Aldrich

3-Nitrobenzyl bromide

98%

Synonym(s):

α-Bromo-m-nitrotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H6NO2Br
CAS Number:
Molecular Weight:
216.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

58-59 °C (lit.)

SMILES string

[O-][N+](=O)c1cccc(CBr)c1

InChI

1S/C7H6BrNO2/c8-5-6-2-1-3-7(4-6)9(10)11/h1-4H,5H2

InChI key

LNWXALCHPJANMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Nitrobenzyl bromide was used in the synthesis of 1,4-disubstituted imidazoles.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D A Heerding et al.
Bioorganic & medicinal chemistry letters, 11(16), 2061-2065 (2001-08-22)
1,4-Disubstituted imidazole inhibitors of Staphylococcus aureus and Escherichia coli enoyl acyl carrier protein reductase (FabI) have been identified. Crystal structure data shows the inhibitor 1 bound in the enzyme active site of E. coli FabI.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service