Skip to Content
Merck
All Photos(1)

Key Documents

218227

Sigma-Aldrich

Cesium hydrogencarbonate

99.9%

Synonym(s):

Cesium bicarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CsHCO3
CAS Number:
Molecular Weight:
193.92
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99.9%

form

solid

SMILES string

[Cs+].OC([O-])=O

InChI

1S/CH2O3.Cs/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

ZMCUDHNSHCRDBT-UHFFFAOYSA-M

General description

Cesium hydrogencarbonate is used as a source of bicarbonate ions.

Application

Cesium hydrogencarbonate can be used as a base:
  • In the Ni-catalyzed hydroarylation of unactivated alkenes with arylboronic acids to synthesize arylated alkanes.
  • To synthesize 5-phenylthiazolamines by coupling thiourea with phenylacetones, phenylacetophenones, and β-tetralone in the presence of CBrCl3.
  • In the Pd-catalyzed hydroxycarbonylation of aryl halides.
  • In the regioselective alkylation of 2,4-dihydroxybenzyaldehydes and 2,4-dihydroxyacetophenones using alkyl halides to synthesize 4-alkylated products.
It can also be used as a catalyst for the:
  • Etherification of glycerol to oligomers.
  • Acetylation of primary alcohols and phenols to acetates using acetic anhydride.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - STOT RE 2 Oral

Target Organs

Kidney,Adrenal gland,Testes

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formulation and utilization of choline based samples for dissolution dynamic nuclear polarization
Bowen S, et al.
Journal of Magnetic Resonance, 236, 26-30 (2013)
Dalton T Snyder et al.
The Analyst, 142(12), 2109-2117 (2017-05-27)
A limitation of conventional quadrupole ion trap scan modes which use rf amplitude control for mass scanning is that, in order to detect a subset of an ion population, the rest of the ion population must also be interrogated. That
Amandine Gendron et al.
Pharmaceutics, 13(1) (2021-01-16)
Cyclosporin A (CsA) is a molecule with well-known immunosuppressive properties. As it also acts on the opening of mitochondrial permeability transition pore (mPTP), CsA has been evaluated for ischemic heart diseases (IHD). However, its distribution throughout the body and its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service