Skip to Content
Merck
All Photos(4)

Key Documents

218170

Sigma-Aldrich

Ethynyltrimethylsilane

98%

Synonym(s):

Trimethylsilylacetylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiC≡CH
CAS Number:
Molecular Weight:
98.22
Beilstein:
906752
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

4.18 psi ( 20 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.388 (lit.)

bp

53 °C (lit.)

density

0.709 g/cm3 (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)C#C

InChI

1S/C5H10Si/c1-5-6(2,3)4/h1H,2-4H3

InChI key

CWMFRHBXRUITQE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Laser-induced polymerization of gaseous ethynyltrimethylsilane was used for efficient chemical vapour deposition of polycarbosilane films. Ethynyltrimethylsilane acts as substrate for nickel-catalyzed cross-coupling with benzonitriles.

Application

Ethynyltrimethylsilane was used in:
  • microwave-assisted, one-pot, three-step Sonogashira cross-coupling-desilylation-cycloaddition reaction for the preparation of 1,4-disubstituted 1,2,3-triazoles
  • synthesis of poly(ethynyltrimethylsilane) containing Pd (II) coordination sites
  • pyrazole synthesis via 1,3-dipolar cycloaddition of diazo compounds to acetylenes

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-29.2 °F

Flash Point(C)

-34 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Shang et al.
Advanced materials (Deerfield Beach, Fla.), 30(27), e1801459-e1801459 (2018-05-26)
In situ weaving an all-carbon graphdiyne coat on a silicon anode is scalably realized under ultralow temperature (25 °C). This economical strategy not only constructs 3D all-carbon mechanical and conductive networks with reasonable voids for the silicon anode at one
Fabian Reiß et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 24(21), 5667-5674 (2018-02-02)
The reaction of the allene precursor Li2 (Me3 SiC3 SiMe3 ) with [Cp2 ZrCl2 ] (Cp=cyclopentadienyl) was examined. The selective formation of hitherto unknown linear, allene-bridged dizirconocene complexes [(Cp2 ZrCl)2 {-μ-(Me3 Si)C3 (SiMe3 )-}] and [(Cp2 Zr)2 {-μ-(Me3 Si)C3 (SiMe3
Atmospheric pressure chemical vapour deposition of polycarbosilane films via UV laser-induced polymerization of ethynyltrimethylsilane.
Pola J, et al.
Surface and Coating Tech., 149(2), 129-134 (2002)
Tetrahedron Letters, 45, 4989-4992 (2004)
Frédéric Friscourt et al.
Organic letters, 12(21), 4936-4939 (2010-10-15)
A microwave-assisted, one-pot, three-step Sonogashira cross-coupling-desilylation-cycloaddition sequence was developed for the convenient preparation of 1,4-disubstituted 1,2,3-triazoles starting from a range of halides, acyl chlorides, ethynyltrimethylsilane, and azides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service