210153
2,2,5-Trimethyl-1,3-dioxane-4,6-dione
97%
Synonym(s):
cycl-Isopropylidene methylmalonate, Methyl Meldrum’s acid, Methylmalonic acid cyclic isopropylidene ester
About This Item
Recommended Products
Assay
97%
form
solid
mp
111-114 °C (lit.)
functional group
ester
SMILES string
CC1C(=O)OC(C)(C)OC1=O
InChI
1S/C7H10O4/c1-4-5(8)10-7(2,3)11-6(4)9/h4H,1-3H3
InChI key
KJMCAXYHYPDRAV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service