Skip to Content
Merck
All Photos(2)

Key Documents

209147

Sigma-Aldrich

Ytterbium(III) nitrate pentahydrate

99.9% trace metals basis

Synonym(s):

Ytterbium trinitrate, Ytterbium(+3) trinitrate pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Yb(NO3)3 · 5H2O
CAS Number:
Molecular Weight:
449.13
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.9% trace metals basis
form:
crystals and lumps
Pricing and availability is not currently available.

Quality Level

Assay

99.9% trace metals basis

form

crystals and lumps

reaction suitability

reagent type: catalyst
core: ytterbium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

SMILES string

O.O.O.O.O.[Yb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/3NO3.5H2O.Yb/c3*2-1(3)4;;;;;;/h;;;5*1H2;/q3*-1;;;;;;+3

InChI key

XIOPWXFTXDPBEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ytterbium(III) nitrate pentahydrate can be used as a precursor to synthesize:
  • Er–Mn: Yb3Al5O12 (YbAG) nanocrystals by nonaqueous sol–gel method. Yb3+–Mn2+ dimer sensitization significantly enhances the green upconversion emissions.
  • CaTiO3:Yb3+, Er3+ nanoparticles by coprecipitation method. These nanoparticles are used in the optical monitoring of mineralization and demineralization of bone scaffolds.
It can also be used as a dopant to prepare BiVO4 semiconductor photocatalysts with enhanced upconversion properties.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nora Jannsen et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(59), 13624-13634 (2019-08-08)
The aim of this work was i) to develop a hydrothermal, low-temperature synthesis protocol affording the upconverting hexagonal phase NaYF4 with suitable dopants while adhering to the "green chemistry" standards and ii) to explore the effect that different parameters have on the

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service