Skip to Content
Merck
All Photos(3)

Key Documents

208477

Sigma-Aldrich

Calcium iodide hydrate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaI2·xH2O
CAS Number:
Molecular Weight:
293.89 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
crystals or chunks

grade

for analytical purposes

Quality Level

Assay

98%

form

crystals or chunks

composition

Degree of hydration, 4-6

mp

783 °C (lit.)

SMILES string

O.I[Ca]I

InChI

1S/Ca.2HI.H2O/h;2*1H;1H2/q+2;;;/p-2

InChI key

XIRQETYMJRSWOH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Compounds with one Saturated Heteroatom Bond-chlorine,bromine,iodine
E.Schaumann
Science of Synthesis: Houben-Weyl Methods of Molecular Transformations, 35 (2006)
Two Carbonylations of Methyl Iodide and Trimethylamine to Acetic acid and N, N-Dimethylacetamide by Rhodium (I) Complex: Stability of Rhodium (I) Complex under Anhydrous Condition.
Hong JH.
Catalysts, 5(4), 1969-1982 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service