Skip to Content
Merck
All Photos(3)

Key Documents

208108

Sigma-Aldrich

Barium titanate(IV)

powder, <3 μm, 99%

Synonym(s):

Titanium barium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaTiO3
CAS Number:
Molecular Weight:
233.19
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

reaction suitability

core: barium
reagent type: catalyst
reagent type: catalyst
core: titanium

particle size

<3 μm

density

6.08 g/mL at 25 °C (lit.)

SMILES string

[Ba++].[O-][Ti]([O-])=O

InChI

1S/Ba.3O.Ti/q+2;;2*-1;

InChI key

WNKMTAQXMLAYHX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Barium titanate(IV) possess piezoelectric, ferroelectric, dielectric and thermoelectric properties. It is a biocompatible ceramic and is therefore used generally in drug delivery.
It has been used:
  • As a precursor to synthesize barium titanate microfibres to be used as building blocks for self-assembled nanoelectronic devices.
  • In multi-material nanoscale 3D printing for developing capacitors, inductors and resistors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of Nanoporous Fibers of BaTiO3 via Electrospinning of BaTiO3/PVC and Thermal Treatment Effects.
Suaste-Gomez E, et al.
Ferroelectrics, 482(1), 1-10 (2015)
Nanoscale 3D printing process using aerodynamically focused nanoparticle (AFN) printing, micro-machining, and focused ion beam (FIB)
Ahn SH, et al.
CIRP Annals, 64(1), 523-526 (2015)
Barium titanate nanoparticles: promising multitasking vectors in nanomedicine.
Genchi GG, et al.
Nanotechnology, 27(23), 232001-232001 (2016)
Experimental studies on 3D printing of barium titanate ceramics for medical applications.
Schult M, et al.
Current Directions in Biomedical Engineering, 2(1), 95-99 (2016)
An efficient approach to derive hydroxyl groups on the surface of barium titanate nanoparticles to improve its chemical modification ability.
Chang SJ, et al.
Journal of Colloid and Interface Science, 329(2), 300-305 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service