Skip to Content
Merck
All Photos(1)

Documents

205613

Sigma-Aldrich

Aliquat® 336

Synonym(s):

Tricaprylylmethylammonium chloride, Trioctylmethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Beilstein:
4039255
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.21

refractive index

n20/D 1.4665 (lit.)

density

0.884 g/mL at 25 °C (lit.)

Application

Mixture of C8 and C10 chains with C8 predominating. Used as a metal extraction reagent and a phase-transfer catalyst.

Legal Information

Aliquat is a registered trademark of BASF

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C - STOT RE 2 Oral

Target Organs

Heart

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

269.6 °F

Flash Point(C)

132 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Proc. Symp. Solvent Extr., 9-9 (1993)
Analyst, 119, 1537-1537 (1994)
Dehmlow, E.V. Dehmlow, S.S.
Phase Transfer Catalysis (Academic Press) (1993)
Alvise Perosa et al.
Chemosphere, 173, 535-541 (2017-02-01)
We describe a chemical technology for the reductive catalytic multiphase hydrodechlorination (HDC) of chlorinated aromatics to greatly reduce their toxicity and aid the disposal of such species. The system requires no solvent and the catalyst displays a high recycling efficiency.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service