Skip to Content
Merck
All Photos(3)

Key Documents

204102

Sigma-Aldrich

Potassium iodide

≥99.99% trace metals basis

Synonym(s):

Knollide, Potassium monoiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystals and lumps

grade

for analytical purposes

vapor pressure

1 mmHg ( 745 °C)

Assay

≥99.99% trace metals basis

form

crystals and lumps

impurities

≤100.0 ppm Trace Metal Analysis

mp

681 °C (lit.)

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium iodide is widely used as an additive to fabricate perovskite solar cells and light-emitting diodes, to enhance their stability and performance. The excess I from KI fill in the Ivacancies and suppress potential pathways for ion migration. The K+ cations can bond with undercoordinated halides or passivate negatively charged trap states at the grain surface, which enhances the performance of the device. It can also be used as a catalyst in various reactions such as the synthesis of biodiesel and cyclic carbonates with five-membered rings.

Application

  • Optimal concentration of potassium iodide to reduce the black staining of silver diamine fluoride: This research identifies the optimal concentration of potassium iodide to minimize staining from silver diamine fluoride in dental applications, relevant for material science focused on dental materials (Detsomboonrat et al., 2022).
  • Optimized assay for hydrogen peroxide determination in plant tissue using potassium iodide: This article discusses an optimized method for determining hydrogen peroxide levels in plant tissues using potassium iodide, useful for academic chemists studying plant physiology or oxidative stress (Junglee et al., 2014).
  • Potassium iodide as a low-cost cathode material for efficient potassium-ion storage: This research investigates the use of potassium iodide as a cost-effective cathode material for potassium-ion batteries, highlighting its potential in energy storage applications (Alanazi et al., 2020).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Maria O'Kane et al.
Nutrients, 10(3) (2018-03-02)
Cow's milk is the most important dietary source of iodine in the UK and Ireland, and also contributes to dietary selenium intakes. The aim of this study was to investigate the effect of season, milk fat class (whole; semi-skimmed; skimmed)
Pietro Santulli et al.
Human reproduction (Oxford, England), 30(1), 49-60 (2014-11-08)
Are protein oxidative stress markers [thiols, advanced oxidation protein products (AOPP), protein carbonyls and nitrates/nitrites] in perioperative peritoneal fluid higher in women with histologically proven endometriosis when compared with endometriosis-free controls? Protein oxidative stress markers are significantly increased in peritoneal
Zhi-Qi Lao et al.
Organic & biomolecular chemistry, 10(39), 7869-7871 (2012-09-06)
We have developed a new KI-catalyzed method for the imidation of an sp(3) C-H bond adjacent to an amide nitrogen atom by using TBHP (tert-butyl hydroperoxide, 70% aqueous solution) as the oxidant. This novel procedure tolerated air and moisture and
Rahul Singh et al.
Carbohydrate polymers, 91(2), 682-685 (2012-11-06)
We report a new biopolymer electrolyte for dye sensitized solar cell application. To develop polymer electrolyte, potassium iodide have been added in agarose biopolymer matrix and characterized using various techniques like complex impedance spectroscopy, Infrared spectroscopy (IR), X-ray diffraction (XRD)
S C Clement et al.
Pediatric blood & cancer, 60(11), 1833-1838 (2013-07-09)
Thyroid dysfunction has been reported in up to 52% of patients 1.4 years after treatment with (131) I-Metaiodobenzylguanidine (MIBG) in children with neuroblastoma (NBL), despite the use of potassium-iodide (KI). Our aim was to investigate if the incidence and severity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service